Would you like to log in again?
PAK2 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-S78S 009-001-S78
75934-078EA 353.41 USD InStock
75934-078 75934-080
PAK2 protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
PAK2 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
PAK2 proteins is a family of serine/threonine kinases that serve as targets for the small GTP binding proteins, CDC42 and RAC1, and have been implicated in a wide range of biological activities. The p21 activated kinases (PAK) are critical effectors that link Rho GTPases to cytoskeleton reorganization and nuclear signaling. PAK2 is activated by proteolytic cleavage during caspase-mediated apoptosis, and may play a role in regulating the apoptotic events in the dying cell (1). The CDC42 and RAC1 induce autophosphorylation of PAK2, which stimulates sustained phosphorylation of other substrates (2). PAK2 Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, Cancer, ERK/MAPK Pathway, Inflammation, Invasion/Metastasis, Neurobiology, and Ser/Thr Kinases research.
PAK2 Protein is stored in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. PAK2 Protein is suitable for use in Western Blot. Expect a band approximately ~87kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty