Would you like to log in again?
PAD4 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-S76S 009-001-S76
75934-066EA 338.75 USD InStock
75934-066 75934-068
PAD4 protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
PAD4 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
PAD4 is a member of the peptidyl arginine deiminase family of enzymes, which catalyze the post-translational deimination of proteins by converting arginine residues into citrullines in the presence of calcium ions (1). PAD4 is essential for antibacterial innate immunity mediated by neutrophil extracellular traps (NETs) (2). PAD4 play a role in granulocyte and macrophage development leading to inflammation and immune response. PAD4 also mediates the gene expression by regulating arginine methylation and citrullination in histones(3). PAD4 Protein is ideal for investigators involved in Signaling Proteins, Arginine Deiminase Proteins, and Inflammation research.
PAD4 Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. PAD4 Protein is suitable for use in Western Blot. Expect a band approximately ~96kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty