p73 beta protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-S71 009-001-S71S
75934-048EA 713.69 USD
75934-048 75934-046
p73 beta protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
p73 beta control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
p73β is a member of the p53 family of transcription factors involved in cellular responses to stress and development. The p73 protein is expressed at very low levels in normal tissues and is differentially expressed in a number of tumors. P73β is strongly involved in malignancy acquisition and maintenance (1). p73 is a stress-response gene that activates transcription of p53-responsive genes and inhibits cell growth in a p53-like manner by inducing apoptosis (2). p73 is a component of a mismatch repair-dependent apoptosis pathway, which contributes to cisplatin-induced cytotoxicity. The regulation of p73 by c-Abl in response to DNA damage was also demonstrated by a failure of ionizing radiation-induced apoptosis after disruption of c-Abl-p73 interaction. p73β Protein is ideal for investigators involved in Signaling Proteins, Cell Cycle Proteins, Apoptosis/Autophagy, Cancer, Cardiovascular Disease, Cell Cycle, Cellular Stress, Inflammation, JNK/SAPK Pathway, Metabolic Disorder, Neurobiology, and p38 Pathway research.
p73β Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. p73β Protein is suitable for use in Western Blot. Expect a band approximately ~88kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.