p70S6KL1 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-S69 009-001-S69S
75930-726EA 364.21 USD
75930-726 75934-040
p70S6KL1 protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
p70S6KL1 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
p70S6KL1 or ribosomal protein S6 kinase-like 1 is a serine/threonine kinase. The p70S6KL1 contains a Microtubule Interacting and Trafficking molecule domain. The p70S6KL1 gene is conserved in chimpanzee, dog, cow, mouse, rat, and chicken. p70S6KL1 Protein is ideal for investigators involved in Signaling Proteins, Cellular Proteins, AKT/PKB Pathway, Angiogenesis, Apoptosis/Autophagy, Cancer, Cardiovascular Disease, Inflammation, Metabolic Disorder, and Ser/Thr Kinases research.
p70S6KL1 Protein is stored in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. p70S6KL1 Protein is suitable for use in Western Blot. Expect a band approximately ~90kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.