Would you like to log in again?
p53 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-S66 009-001-S66S
75930-708EA 665.18 USD InStock
75930-708 75930-704
p53 protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
p53 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
p53 is a key regulator of cell growth and acts as a tumor suppressor gene. Wild-type p53 gene can suppress transformation of rat embryo fibroblasts in cell culture by other oncogenes such as the adenovirus E1A and Ras (1). DNA tumor viruses such as SV40 large T antigen and the adenovirus E1A plus E1B-55Kd proteins bind to p53 and inactivate its tumor suppressor activities leading to cellular transformation. Mutational inactivation of the p53 gene is detected in more than 50% of human cancers. Mutation of p53 renders cancer cells more resistant to current cancer therapies due to lack of p53-mediated apoptosis (2). p53 Protein is ideal for investigators involved in Signaling Proteins, Cell Cycle Proteins, Apoptosis/Autophagy, Cancer, Cardiovascular Disease, Cell Cycle, Cellular Stress, Inflammation, JNK/SAPK Pathway, Metabolic Disorder, Neurobiology, and p38 Pathway research.
p53 Protein is stored in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 0.25mM DTT, 0.1mM EDTA, 0.1mM PMSF, 10mM glutathione, and 25% glycerol. p53 Protein is suitable for use in Western Blot and Kinase Assay. Expect a band approximately ~79kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty