Would you like to log in again?
p33ING1 protein-HIS Epitope, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-S64S 009-001-S64
75930-692EA 338.75 USD InStock
75930-692 75930-694
p33ING1 protein-HIS Epitope, Rockland Immunochemicals
Proteins and Peptides
p33ING1 control protein-HIS Epitope
  • Control Protein
  • PCA Assays
  • ImmunoBlot
p33ING1 displays characteristics of a tumor suppressor protein and can induce cell growth arrest and apoptosis. p33ING1 is a nuclear protein and acute expression of p33 ING1 inhibits cell growth. p33ING1 physically interacts with the tumor suppressor protein TP53 and is a component of the p53 signaling pathway. Mutation of p33ING1 gene occurs in neuroblastoma cells and reduced expression is observed in some breast cancer cell lines (1). It has been proposed that p33ING1 can act as a potent growth regulator in normal and in established cells and plays a role as a candidate tumor suppressor gene whose inactivation may contribute to the development of cancers (2). p33ING1 Protein is ideal for investigators involved in Signaling Proteins, Cell Cycle Proteins, Cancer, and Cell Cycle research.
p33ING1 Protein is stored in 50mM MOPS, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol. p33ING1 Protein is suitable for use in Western Blot. Expect a band approximately ~38kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty