Would you like to log in again?
p18INK4C protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-S59 009-001-S59S
75930-670EA 665.18 USD InStock
75930-670 75930-668
p18INK4C protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
p18INK4C control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
p18INK4C is a member of the INK4 family of proteins that regulate the G1 to S cell cycle transition by binding to and inhibiting the pRb kinase activity of cyclin-dependent kinases 4 and 6 (1). Mutation of p18INK4C impairs B-cell terminal differentiation and confers increased susceptibility to tumor development. p18INK4C can function as a tumor suppressor gene in Hodgkins lymphoma and its inactivation may contribute to the cell cycle deregulation and defective terminal differentiation characteristic of the Reed-Sternberg cells (2). p18INK4C Protein is ideal for investigators involved in Signaling Proteins, Cell Cycle Proteins, Cancer, and Cell Cycle research.
p18INK4C Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. p18INK4C Protein is suitable for use in Western Blot and Kinase Assay. Expect a band approximately ~42kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty