Would you like to log in again?
NFKB2 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-S57 009-001-S57S
75930-416EA 665.18 USD InStock
75930-416 75930-414
NFKB2 protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
NFKB2 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
NFKB2 is a part of the NFKB complex that is present in various cell types that express cytokines, chemokines, growth factors, cell adhesion molecules, and some acute phase proteins in health and disease states. NFKB protein is activated by a wide variety of stimuli such as cytokines, oxidant-free radicals, inhaled particles, ultraviolet irradiation, and bacterial or viral products (1). Inappropriate activation of NFKB has been linked to inflammatory events associated with autoimmune arthritis, asthma, septic shock, lung fibrosis, glomerulonephritis, atherosclerosis, and AIDS (2). Overexpression of NFKB2 protects androgen sensitive LNCaP cells from apoptotic cell death and cell cycle arrest induced by androgen-deprivation. NFKB2 Protein is ideal for investigators involved in Signaling Proteins, Transcription Proteins, AKT/PKB Pathway, Apoptosis/Autophagy, Inflammation, and p38 Pathway research.
NFKB2 Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. NFKB2 Protein is suitable for use in Western Blot. Expect a band approximately ~84kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty