Would you like to log in again?
MGRN1 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-S48 009-001-S48S
75934-878EA 665.18 USD InStock
75934-878 75934-876
MGRN1 protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
MGRN1 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
MGRN1 or mahogunin ring finger 1, E3 ubiquitin protein ligase is a C3HC4 RING-containing protein with E3 ubiquitin ligase activity in vitro (1). MGRN1 gene encodes 4 mRNA isoforms that contain a central C3HC4 RING domain as the only recognizable protein motif and regions flanking the RING domain and the N-terminal region of the protein are also conserved between vertebrate and invertebrate genome and MGRN1 is highly expressed in brain (2). MGRN1 has been detected in the adrenal cortex in kidney and its role may relate to the trafficking and/or degradation of the melanocortin 2 receptor. MGRN1 Protein is ideal for investigators involved in Signaling Proteins, Ubiquitin Proteins, Angiogenesis, Apoptosis/Autophagy, Cancer, Cell Cycle, Cellular Stress, Inflammation, Invasion/Metastasis, and Neurobiology research.
MGRN1 Protein is stored in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. MGRN1 Protein is suitable for use in Western Blot and Kinase Assay. Expect a band approximately ~96kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty