Would you like to log in again?
KDM1A protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-S31S 009-001-S31
75933-786EA 353.41 USD InStock
75933-786 75933-788
KDM1A protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
KDM1A control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
KDM1A or lysine (K)-specific demethylase 1A is a nuclear protein containing a SWIRM domain, a FAD-binding motif, and an amine oxidase domain that is a component of several histone deacetylase complexes and act as a histone demethylase. (1) H3K4 histone demethylase activity of KDM1A is partly responsible for the repressive activity of TAL1 and restricts TAL1 function in hematopoiesis (1). KDM1A plays an essential role for CoREST in demethylation of H3K4 both in vitro and in vivo (2). KDM1A Protein is ideal for investigators involved in Signaling Proteins, Deacetylase/Demethylase Proteins, Cancer, Cell Cycle, and Inflammation research.
KDM1A Protein is stored in 50mM Tris-HCl, pH 7.5, 50mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. KDM1A Protein is suitable for use in Western Blot. Expect a band approximately ~100kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty