Would you like to log in again?
KAT8 (MYST1) protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-S29S 009-001-S29
75929-402 75929-404
KAT8 (MYST1) protein-GST fusion, Rockland Immunochemicals
KAT8 (MYST1) control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
KAT8 (MYST1) or MYST histone acetyltransferase 1 is a signaling protein that belongs to the MYST family of histone acetyl transferases (HATs) and was originally isolated as an HIV-1 TAT-interactive protein which play important roles in regulating chromatin remodeling, transcription and other nuclear processes by acetylating histone and nonhistone proteins. MYST1 have a chromodomain that involved in protein-protein interactions and targeting transcriptional regulators to chromatin (1). MYST histone acetyltransferase 1 is also involved in ATM function (2). KAT8 Protein is ideal for investigators involved in Signaling Proteins, Acetyl/Methyltransferase Proteins, Apoptosis/Autophagy, Cancer, Cardiovascular Disease, Cell Cycle, ERK/MAPK Pathway, Inflammation, Invasion/Metastasis, Metabolic Disorder, Neurobiology, NfkB Pathway, and PKA/PKC Pathway research.
KAT8 Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. KAT8 Protein is suitable for use in Western Blot. Expect a band approximately ~83kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty