Would you like to log in again?
I kappaB alpha protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-S17 009-001-S17S
75928-842EA 627.23 USD InStock
75928-842 75928-838
I kappaB alpha protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
I kappaB alpha control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
IkB-alpha is an inhibitor of the NFkB complex and inactivates the NFkB by trapping it in the cytoplasm (1). Phosphorylation of serine residues on the IkB protein by IkB kinases (IKKs) marks it for destruction via the ubiquitination pathway, thereby allowing the activation of the NFkB complex. Synthetic glucocorticoid such as dexamethasone display anti-inflammatory effects by inducing the increased synthesis of the IkB protein thereby inhibiting the activity of the NFkB complex. Overexpression of the IkB-alpha gene in fibroblasts leads to inhibition of production of IL-6, TNF receptor, MMP-1, MMP-3 and MMP-13 (2). IkB-alpha Protein is ideal for investigators involved in Signaling Proteins, Transcription Proteins, AKT/PKB Pathway, Apoptosis/Autophagy, Inflammation, and p38 Pathway research.
IkB-alpha Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. IkBα Protein is suitable for use in Western Blot and Kinase Assay. Expect a band approximately ~61kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty