HSPA1AB
Supplier: Rockland Immunochemicals
009-001-S14S 009-001-S14
75928-750EA 713.69 USD
75928-750 75928-746
HSPA1AB
Proteins and Peptides
HSP70 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
HSP70 is a member of the heat shock protein family and is synthesized by cells of many organisms in response to stress (1). HSP70 is found mostly but not exclusively in the nucleus of unstressed cells. For several hours after a short heat shock, however, it is strongly concentrated in nucleoli (2). Nucleoli are transiently damaged by such a heat shock: their morphology changes and assembly and export of ribosomes is blocked for several hours. HSP70 helps to stabilize the morphological changes of the nucleoli. HSP70 Protein is ideal for investigators involved in Signaling Proteins, Cell Stress & Chaperone Proteins, Apoptosis/Autophagy, Cancer, and Cellular Stress research.
HSP70 Protein is stored in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol. HSP70 Protein is suitable for use in Western Blot and ATPase Assay. Expect a band approximately ~70kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.