Would you like to log in again?
HSP10 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-S09S 009-001-S09
75928-736EA 353.41 USD InStock
75928-736 75928-738
HSP10 protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
HSP10 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
HSP10 is a 10-kDa stress-inducible mitochondrial protein which is a member of the heat shock proteins. HSP10 is synthesized at elevated rates in cultured rat hepatoma cells challenged with heat shock or amino acid analogues. HSP10 has been implicated in multiple disease states in humans. In myocardial samples from patients with chronic atrial fibrillation, cardiomyocytes respond to chronic atrial fibrillation with increased expression of HSP10 and HSP60 (1). HSP10 is also overexpressed early in prostate carcinogenesis. HSP10 is also overexpressed in large bowel carcinomas and it may have diagnostic and prognostic significance in the management of this tumour (2). HSP10 Protein is ideal for investigators involved in Signaling Proteins, Cell Stress & Chaperone Proteins, Cardiovascular Disease, and Cellular Stress research.
HSP10 Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. HSP10 Protein is suitable for use in Western Blot. Expect a band approximately ~36kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty