Would you like to log in again?
HO1 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-S03S 009-001-S03
75928-504EA 353.41 USD InStock
75928-504 75928-506
HO1 protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
HO1 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
HO1 (Heme oxygenase 1) catalyzes the oxidative cleavage of heme to biliverdin and is one of the main genes controlling heme synthesis and catabolism. HO1 plays a protective role in various disorders and can ameliorate experimental MN via multiple pathways, including anti-oxidative and immunomodulatory effects (1). Exposure of primary hepatocytes to carbon monoxide and nitric oxide results in dramatic induction of HO1 in dose- and time-dependent manner and this induction is blocked by MAP kinase inhibitors (MAPKs) but not by inhibitors of PI3 kinase pathway (2). HO1 Protein is ideal for investigators involved in Signaling Proteins, Cell Stress & Chaperone Proteins, Cancer, Cardiovascular Disease, Cellular Stress, Inflammation, and Neurobiology research.
HO1 Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. HO1 Protein is suitable for use in Western Blot. Expect a band approximately ~56kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty