Would you like to log in again?
GRB2 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-R99S 009-001-R99
75934-454EA 353.41 USD InStock
75934-454 75934-456
GRB2 protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
GRB2 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
GRB2 is an adaptor protein that is the homolog of the Sem5 gene of C. elegans and Drk gene of Drosophila. GRB2 protein contains one SH2 domain and two SH3 domains and plays a role in growth factor-mediated signal transduction. GRB2 forms a complex with activated EGFR and the RAS-specific guanine nucleotide exchange factor SOS1 thereby mediating the growth factor-induced activation of RAS (1). GRB2 has been implicated in growth factor regulation of the cytoskeleton and DNA synthesis. DAB2 also binds to GRB2 and competes with SOS for GRB2 binding. GRB2 interacts with the hepatitis C virus nonstructural 5A (NS5A) protein and inhibits the activation of ERK1 and ERK2 by EGF (2). GRB2 Protein is ideal for investigators involved in Signaling Proteins, Adaptor Proteins, Angiogenesis, Apoptosis/Autophagy, Cancer, Cardiovascular Disease, ERK/MAPK Pathway, Invasion/Metastasis, JAK/STAT Pathway, and Neurobiology research.
GRB2 Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. GRB2 Protein is suitable for use in Western Blot. Expect a band approximately ~49kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty