Would you like to log in again?
Human Fractalkine (CX3CL1) Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009001H49-0100
10801-480EA 565.01 USD InStock
10801-480
Human Fractalkine (CX3CL1) Protein, Rockland Immunochemicals
Proteins and Peptides
rHuman Fractalkine (CX3CL1) control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Fractalkine, also known as CX3CL1, is an atypical chemokine that was the first of a fourth chemokine motif (CX3C). It is thought to function as a T cell and monocyte chemotractant and is produced by non-haemopoietic cells. Fractalkine is made in a soluble and membrane bound form in activated endothelial cells which is thought to promote adhesion of leukocytes. Recombinant human Fractalkine is a non-glycosylated protein, containing 76 amino acids, with a molecular weight of 8.6 kDa.
Fractalkine Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Fractalkine in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty