Would you like to log in again?
ERK2 double mutant hu recomb protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-GR2
76221-878EA 337.65 USD InStock
76221-878
ERK2 double mutant hu recomb protein, Rockland Immunochemicals
Proteins and Peptides
ERK2 double mutant human recombinant control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
ERK2 isoform double mutant recombinant protein is modified to put alanine residues at two key acitivation sites, Threonine 185 and Tyrosine 222. Mitogen activiated protein kinase 1, also known as MAPK1, ERK, or ERK2, is an integral component of the MAP kinase cascade that regulates cell growth and differentiation. This pathway also plays a key role in synaptic plasticity in the brain. The inactive double mutant ERK2 recombinant protein is ideal for investigators involved in Neuroscience, Cell Signaling and Cancer Research.
ERK2 mutant protein is suitable as a negative control protein for immunoassays using antibodies targeting the critical ERK2 phosphorylated T185 or Y187 residues. For western blot use at 50 ng or less. For other assays concentration is user optimized.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty