ERK1 double mutant human recombinant control protein
- Control Protein
- PCA Assays
- ImmunoBlot
ERK1 isoform double mutant recombinant protein is modified to put alanine residues at two key acitivation sites, Threonine 202 and Tyrosine 204. Mitogen activiated protein kinase 3, also known as MAPK3, ERK, or ERK1, is an integral component of the MAP kinase cascade that regulates cell growth and differentiation. This pathway also plays a key role in synaptic plasticity in the brain. The inactive double mutant ERK1 recombinant protein is ideal for investigators involved in Neuroscience, Cell Signaling and Cancer Research.
ERK1 mutant protein is suitable as a negative control protein for immunoassays using antibodies targeting the critical ERK1 phosphorylated T202 or Y204 residues. For western blot use at 50 ng or less. For other assays concentration is user optimized.