Would you like to log in again?
ERK2 pT185-pY187 hu recomb protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-GR0
76221-874EA 337.65 USD InStock
76221-874
ERK2 pT185-pY187 hu recomb protein, Rockland Immunochemicals
Proteins and Peptides
ERK2 pT185-pY187 human recombinant control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
ERK2 pT185-pY187 activated recombinant protein was produced in Sf9 cells. Mitogen activiated protein kinase 1, also known as MAPK1, ERK, or ERK2, is an integral component of the MAP kinase cascade that regulates cell growth and differentiation. This pathway also plays a key role in synaptic plasticity in the brain. This recombninant protein is the activated ERK1 isoform and is ideal for investigators involved in Neuroscience, Cell Signaling and Cancer Research.
ERK2 pT185-pY187 is an activated recombinant protein suitable as a control for polyclonal or monoclonal anti-ERK in immunological assays. ERK2 can be activated by in vitro phosphorylation by MEK proteins. For western blot use at 50 ng or less. For other assays concentration is user optimized.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty