Would you like to log in again?
MEK1 pS218-pS222 hu recomb protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-GP5
76221-856EA 337.65 USD InStock
76221-856
MEK1 pS218-pS222 hu recomb protein, Rockland Immunochemicals
Proteins and Peptides
MEK1 pS218-pS222 human recombinant control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
MEK1 pS218-pS222 is an activated recombinant protein was produced in Sf9 cells by co-expression of B-raf. Mitogen-activated protein kinase kinase 1, also known as MKK or MEK, is an integral component of the MAP kinase cascade that regulates cell growth and differentiation. This pathway also plays a key role in synaptic plasticity in the brain. Activated MEK 1 acts as a dual specificity kinase phosphorylating both a threonine and a tyrosine residue on MAP kinase. This recombninat protein is the MEK1 isoform and is ideal for investigators involved in Neuroscience, Cell Signaling and Cancer Research.
MEK1 pS218-pS222 is an activated recombinant protein suitable as a control for polyclonal or monoclonal anti-MEK1 in immunological assays. Activated MEK1 can be used in vitro to phosphorylate ERK protein. For western blot use at 50 ng or less. For other assays concentration is user optimized.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty