Would you like to log in again?
HSF1 protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-F50 009-001-F50S
75928-732EA 665.18 USD InStock
75928-732 75928-730
HSF1 protein, Rockland Immunochemicals
Proteins and Peptides
HSF1 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
HSF1 is a member of the heat shock transcription factor family. Protein-damaging stress lead to the activation of HSF1 which binds to upstream regulatory sequences in the promoters of heat shock genes leading to enhanced heat shock gene expression (1). The activation of HSF1 proceeds through a multi-step pathway, involving a monomer-to-trimer transition, nuclear accumulation and extensive posttranslational modifications. HSF1 activity is regulated at different levels by heat shock proteins and co-chaperones and is modulated further by a number of mechanisms involving other stress-regulated aspects of cell metabolism (2). HSF1 Protein is ideal for investigators involved in Signaling Proteins, Transcription Proteins, Cancer, Cardiovascular Disease, Cellular Stress, Inflammation, and JNK/SAPK Pathway research.
HSF1 Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. HSF1 Protein is suitable for use in Western Blot and Kinase Assay. Expect a band approximately ~96kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty