Would you like to log in again?
EGF Human Recombinant Epidermal Growth Factor, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-C26
RL009001C26EA 311.76 USD InStock
RL009001C26
EGF Human Recombinant Epidermal Growth Factor, Rockland Immunochemicals
Proteins and Peptides
EGF Human Recombinant Epidermal Growth Factor control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
EGF (Epidermal Growth Factor) protein, of the superfamily with the same name, is synthesized as a large precursor molecule that is proteolytically cleaved to generate the 53-amino acid epidermal growth factor peptide. This protein acts a potent mitogenic factor that plays an important role in the growth, proliferation and differentiation of numerous cell types. It is a magnesiotropic hormone that stimulates magnesium reabsorption in the renal distal convoluted tubule via engagement of EGFR and activation of the magnesium channel TRPM6. This protein acts by binding the high affinity cell surface receptor, epidermal growth factor receptor. Defects in this gene are the cause of hypomagnesemia type 4 (HOMG4). Dysregulation of this gene has been associated with the growth and progression of certain cancers. Alternate splicing results in multiple transcript variants. This protein exists in the membrane as a single-pass type I membrane protein, and contains 9 EGF-like domains as well as 9 LDL-receptor class B repeats. It is significantly expressed in kidney, salivary gland, cerebrum and prostate.
EGF Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-EGF in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty