Would you like to log in again?
IL-29 Human Recombinant Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-C24
RL009-001-C24EA 311.76 USD InStock
RL009-001-C24
IL-29 Human Recombinant Protein, Rockland Immunochemicals
Proteins and Peptides
IL-29 Human Recombinant control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
IL-29 (Interleukin-29) is a secreted cytokine with immunomodulatory activity. It may play a role in antiviral immunity. It up-regulates MHC class I antigen expression. IL-29 is a ligand for the heterodimeric class II cytokine receptor composed of IL10RB and IL28RA. The ligand/receptor complex seems to signal through the Jak-STAT pathway. Its induction is by viral infections or double stranded RNA. Recombinant IL-29 produced in E.coli is a single, non-glycosylated polypeptide chain containing 179 amino acids and having a molecular mass of 19,895 Daltons.
IL-29 is suitable as a control for polyclonal or monoclonal anti-IL-29 in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty