Would you like to log in again?
MIP-3ß Human Recombinant Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-C21
RL009-001-C21EA 311.76 USD InStock
RL009-001-C21
MIP-3ß Human Recombinant Protein, Rockland Immunochemicals
Proteins and Peptides
MIP-3ß Human Recombinant control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
The MIP-3ß (also known as SCYA19) member of the intercrine beta (chemokine CC) family may play a role not only in inflammatory and immunological responses, but also in normal lymphocyte recirculation and homing. May play an important role in trafficking of T-cells in thymus, and T-cell and B-cell migration to secondary lymphoid organs. It is secreted. It specifically binds to chemokine receptor CCR7. MIP3ß is expressed at high levels in the lymph nodes, thymus and appendix, and at intermediate levels seen in colon and trachea, while low levels found in spleen, small intestine, lung, kidney and stomach. Recombinant Human MIP-3b produced in E. coli is a single, non-glycosylated, polypeptide chain containing 77 amino acids and having a molecular mass of 8809 Daltons. The recombinant protein shows potent chemotactic activity for T-cells and B-cells but not for granulocytes and monocytes.
MIP-3ß Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-MIP-3ß in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty