Would you like to log in again?
Human RANTES (CCL5) Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009001C20-0100
75931-446
Human RANTES (CCL5) Protein, Rockland Immunochemicals
rHuman RANTES (CCL5) control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Regulated Upon Activation Normal T cell Express Sequence (RANTES), also called CCL5, is a chemokine produced by T cells three to five days after activation. RANTES is a promiscuous chemokine that signals through several G protein-coupled receptors, CCR5, CCR3, CCR1 and US28 (a viral receptor encoded by human CMV). The main function of RANTES is to recruit immune cells to the site of inflammation. Recombinant human RANTES is a non-glycosylated protein, containing 68 amino acids, with a molecular weight of 7.8 kDa.
RANTES Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-RANTES in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty