Would you like to log in again?
IL-3 Recombinant Human Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001B92-0.1
RL009001B920.1
IL-3 Recombinant Human Protein, Rockland Immunochemicals
IL-3 Recombinant Human control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Interleukin-3 (IL-3) is a cytokine produced by activated T-cells and mast cells that is active on a broad range of hematopoietic cells and in the nervous system and appears to be important in several chronic inflammatory diseases. Recombinant Human IL-3 produced in E. coli is a single, non-glycosylated polypeptide chain containing 133 amino acids and having a molecular mass of 15,000 Daltons.
IL-3 is suitable as a control for polyclonal or monoclonal anti-IL-3 in immunological assays. Buffer formation: 10 mM sodium phosphate, 50 mM sodium sulfate, pH 7.5.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty