Would you like to log in again?
MIP-3α Human Recombinant Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-B51
RL009-001-B51EA 311.76 USD InStock
RL009-001-B51
MIP-3α Human Recombinant Protein, Rockland Immunochemicals
Proteins and Peptides
MIP-3α Human Recombinant control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Human Macrophage Inflammatory Protein-3 alpha, also known as LARC (Liver and Activation Regulated Chemokine) or CCL20. This protein is strongly up regulated by inflammatory signals and down regulated by the anti-inflammatory cytokine IL-10. H-MIP-3-alpha is a non-glycosylated polypeptide chain consisting of 70 amino acids and a molecular weight of 8.0 kDa.
MIP-3α is suitable as a control for polyclonal or monoclonal anti-MIP-3α in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty