Would you like to log in again?
IL-32A Human Recombinant Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-B49
RL009-001-B49EA 311.76 USD InStock
RL009-001-B49
IL-32A Human Recombinant Protein, Rockland Immunochemicals
Proteins and Peptides
IL-32A Human Recombinant control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Human IL-32 alpha is one of approximately 6 splice variants of a gene cloned from the human lung carcinoma stable transfectant, A549-Rbeta. IL-32 alpha has been shown to induce IL-8, TNF-a, and MIP-2 production from human & mouse macrophage cell lines. It is upregulated in activated T- & NK-cells, and IFN-g-treated epithelial cells. Human recombinant IL-32 alpha produced in E. coli is a 131 amino acid non -glycosylated polypeptide with a molecular mass of 14.9 kDa.
IL-32A is suitable as a control for polyclonal or monoclonal anti-IL-32A in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty