Would you like to log in again?
MIP-1α Human Recombinant Protein, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-315
RL009-001-315
MIP-1α Human Recombinant Protein, Rockland Immunochemicals
MIP-1α Human Recombinant control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Recombinant Human Macrophage Inflammatory Protein-1 (CCL3, MIP-1) produced in E. coli is a single, non-glycosylated, polypeptide chain containing 70 amino acids and having a molecular mass of 7820 Daltons.
MIP-1α is suitable as a control for polyclonal or monoclonal anti--MIP-1α in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty