p53 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-174S 009-001-174
75930-702EA 364.21 USD
75930-702 75930-706
p53 protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
p53 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
p53 is a key regulator of cell growth and acts as a tumor suppressor gene. Wild-type p53 gene can suppress transformation of rat embryo fibroblasts in cell culture by other oncogenes such as the adenovirus E1A and Ras (1). DNA tumor viruses such as SV40 large T antigen and the adenovirus E1A plus E1B-55Kd proteins bind to p53 and inactivate its tumor suppressor activities leading to cellular transformation. Mutational inactivation of the p53 gene is detected in more than 50% of human cancers. Mutation of p53 renders cancer cells more resistant to current cancer therapies due to lack of p53-mediated apoptosis (2). p53 Protein is ideal for investigators involved in Signaling Proteins, Cell Cycle Proteins, Apoptosis/Autophagy, Cancer, Cardiovascular Disease, Cell Cycle, Cellular Stress, Inflammation, JNK/SAPK Pathway, Metabolic Disorder, Neurobiology, and p38 Pathway research.
p53 Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.25mM DTT, 0.1mM EGTA, 0.1mM EDTA, 0.1mM PMSF, 25% glycerol. p53 Protein is suitable for use in Western Blot and Kinase Assay. Expect a band approximately ~79kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.