Would you like to log in again?
p21CIP1 protein-GST fusion, Rockland Immunochemicals
Supplier: Rockland Immunochemical
009-001-171S 009-001-171
75930-676EA 353.41 USD InStock
75930-676 75930-678
p21CIP1 protein-GST fusion, Rockland Immunochemicals
Proteins and Peptides
p21CIP1 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
CIP 1 (Cyclin-Dependent Kinase Inhibitor 1A) regulates cell cycle progression, terminal differentiation, and apoptosis (1). CIP1 was shown to be induced by p53 and to be a potent inhibitor of cyclin-dependent kinase (CDK) activity. DNA damage leads to increased expression of CIP1 in cyclin E-containing complexes and to an associated decrease in cyclin-dependent kinase activity. CIP1 is a critical downstream effector in the p53-specific pathway of growth control in mammalian cells (2). p21CIP1 Protein is ideal for investigators involved in Signaling Proteins, Cell Cycle Proteins, Cancer, and Cell Cycle research.
p21CIP1 Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. p21CIP1 Protein is suitable for use in Western Blot and Kinase Assay. Expect a band approximately ~46kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty