Would you like to log in again?
Horse Albumin, Rockland Immunochemicals
Supplier: Rockland Immunochemical
008-0133-0010
75928-512
Horse Albumin, Rockland Immunochemicals
Horse Albumin
  • protein standard
Albumins are a group of water soluble blood proteins that are non-glycosylated and are capable of heat denaturation. Many albumins act as transport proteins in the blood, binding to water, cations (such as calcium, sodium, and potassium), lipids, as well as some hormones and drugs.
Horse Albumin is ideal for investigators in Immunology, Cancer, and Microbiology research.
Horse Albumin can be utilized as a protein standard in ELISA, Immunohistochemistry, Western Blot experiments, and as a nutrient for cell culture.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty