Horse IgG
Supplier: Rockland Immunochemicals
008-0102-0010
75934-474EA 163.13 USD
75934-474
Horse IgG
Proteins and Peptides
Horse IgG Whole Molecule
  • control or standard reagent
Secreted as part of the adaptive immune response by plasma B cells, immunoglobulin G constitutes 75% of serum immunoglobulins. Immunoglobulin G binds to viruses, bacteria, as well as fungi and facilitates their destruction or neutralization via agglutination (and thereby immobilizing them), activation of the compliment cascade, and opsinization for phagocytosis. The whole IgG molecule possesses both the F(c) region, recognized by high-afinity Fc receptor proteins, as well as the F(ab) region possessing the epitope-recognition site. Both heavy and light chains of the antibody molecule are present.
House IgG whole molecule can be utilized as a control or standard reagent in Western Blotting and ELISA experiments.