Guinea Pig IgG
Supplier: Rockland Immunochemicals
006-0102-0005
75928-304EA 238.24 USD
75928-304
Guinea Pig IgG
Antibodies
Guinea Pig IgG Whole Molecule
  • control or standard reagent
Secreted as part of the adaptive immune response by plasma B cells, immunoglobulin G constitutes 75% of serum immunoglobulins. Immunoglobulin G binds to viruses, bacteria, as well as fungi and facilitates their destruction or neutralization via agglutination (and thereby immobilizing them), activation of the compliment cascade, and opsinization for phagocytosis. The whole IgG molecule possesses both the F(c) region, recognized by high-afinity Fc receptor proteins, as well as the F(ab) region possessing the epitope-recognition site. Both heavy and light chains of the antibody molecule are present.
Guinea Pig IgG whole molecule can be utilized as a control or standard reagent in Western Blotting and ELISA experiments.