Goat IgG (AP (Alkaline Phosphatase))
Supplier: Rockland Immunochemicals
005-0502
RL005-0502EA 176.9 USD
RL005-0502
Goat IgG (AP (Alkaline Phosphatase))
Antibodies
Goat IgG Whole Molecule Alkaline Phosphatase
  • control or standard reagent
Secreted as part of the adaptive immune response by plasma B cells, immunoglobulin G constitutes 75% of serum immunoglobulins. Immunoglobulin G binds to viruses, bacteria, as well as fungi and facilitates their destruction or neutralization via agglutination (and thereby immobilizing them), activation of the compliment cascade, and opsinization for phagocytosis. The whole IgG molecule possesses both the F(c) region, recognized by high-affinity Fc receptor proteins, as well as the F(ab) region possessing the epitope-recognition site. Both heavy and light chains of the antibody molecule are present.
Goat IgG whole molecule alkaline phosphatase conjugate can be utilized as a control or standard reagent in Western Blotting and ELISA experiments.