Goat IgG Fab
Supplier: Rockland Immunochemicals
005-0105-0001
10801-802EA 301.94 USD
10801-802
Goat IgG Fab
Antibodies
Goat IgG Fab Fragment
  • control or standard reagent
Secreted as part of the adaptive immune response by plasma B cells, immunoglobulin G constitutes 75% of serum immunoglobulins. Immunoglobulin G binds to viruses, bacteria, as well as fungi and facilitates their destruction or neutralization via agglutination (and thereby immobilizing them), activation of the compliment cascade, and opsinization for phagocytosis. This product possesses the F(ab) region possessing the epitope-recognition site, both heavy and light chains of the antibody molecule are present.
Goat IgG F(ab) can be utilized as a control or standard reagent in Western Blotting and ELISA experiments.