Would you like to log in again?
Goat IgG F(ab')2, Rockland Immunochemicals
Supplier: Rockland Immunochemical
005-0104-0001
10801-784
Goat IgG F(ab')2, Rockland Immunochemicals
Goat IgG F(ab')2 Fragment
  • control or standard reagent
Secreted as part of the adaptive immune response by plasma B cells, immunoglobulin G constitutes 75% of serum immunoglobulins. Immunoglobulin G binds to viruses, bacteria, as well as fungi and facilitates their destruction or neutralization via agglutination (and thereby immobilizing them), activation of the compliment cascade, and opsinization for phagocytosis. This product possesses the F(ab')2 fragment, recognized by the two F(ab) fragments yielded from the digestion of the antibody below the disulfide bond hinge region.
Goat IgG F(ab')2 fragment can be utilized as a control or standard reagent in Western Blotting and ELISA experiments.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty