Dog IgG F(ab')2 Fragment
- control or standard reagent
Secreted as part of the adaptive immune response by plasma B cells, immunoglobulin G constitutes 75% of serum immunoglobulins. Immunoglobulin G binds to viruses, bacteria, as well as fungi and facilitates their destruction or neutralization via agglutination (and thereby immobilizing them), activation of the compliment cascade, and opsinization for phagocytosis.
This product possesses the F(ab')2 fragment, recognized by the two F(ab) fragments yielded from the digestion of the antibody below the disulfide bond hinge region.
Dog IgG F(ab')2 fragment can be utilized as a control or standard reagent in Western Blotting and ELISA experiments.