Bovine IgG (AP (Alkaline Phosphatase))
Supplier: Rockland Immunochemicals
001-0502
RL001-0502EA 176.93 USD
RL001-0502
Bovine IgG (AP (Alkaline Phosphatase))
Antibodies
Bovine IgG whole molecule Alkaline Phosphatase
  • control or standard reagent
Secreted as part of the adaptive immune response by plasma B cells, immunoglobulin G constitutes 75% of serum immunoglobulins.
Immunoglobulin G binds to viruses, bacteria, as well as fungi and facilitates their destruction or neutralization via agglutination (and thereby immobilizing them), activation of the compliment cascade, and opsinization for phagocytosis.
The whole IgG molecule possesses both the F(c) region, recognized by high-affinity Fc receptor proteins, as well as the F(ab) region possessing the epitope-recognition site. Both heavy and light chains of the antibody molecule are present.
Bovine IgG whole molecule alkaline phosphatase conjugation can be utilized as a control or standard reagent in Western Blotting and ELISA experiments.