Would you like to log in again?
Histatin 5, Rockland Immunochemicals
Supplier: Rockland Immunochemical
000-001-M59
RL000-001-M59EA 275.74 USD InStock
RL000-001-M59
Histatin 5, Rockland Immunochemicals
Proteins and Peptides
Histatin Control Peptide
  • Control Peptide - Blocking Peptide
  • PCA Assays
  • ImmunoBlot
Histatins are antimicrobial and antifungal proteins found in the saliva. They play a role in wound closure. The three major histatins are HTN1, HTN3, and HTN5.
Histatin 5 Control Peptide is suitable for use in ELISA, Western Blot, Dot blot, PCA, and other assays. Control peptide should be used at 1.0 µg per 1.0 µl of antiserum in per assay. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty