hObestatin, Rockland Immunochemicals
Supplier: Rockland Immunochemical
000-001-M50
RL000-001-M50EA 291.09 USD
RL000-001-M50
hObestatin, Rockland Immunochemicals
Proteins and Peptides
Human Obestatin Control Peptide
  • Control Peptide - Blocking Peptide
  • PCA Assays
  • ImmunoBlot
Obestatin, a putative peptide hormone, is a small protein encoded by the same gene that also encodes ghrelin, the peptide hormone that increases appetite.
Obestatin Control Peptide is suitable for use in ELISA, Western Blot, Dot blot, PCA, and other assays. Control peptide should be used at 1.0 µg per 1.0 µl of antiserum in per assay. Specific conditions for reactivity should be optimized by the end user.