Ovalbumin 257-264
Supplier: Rockland Immunochemicals
000-001-M45
RL000-001-M45EA 91.39 USD
RL000-001-M45
Ovalbumin 257-264
Proteins and Peptides
Ovalbumin Control Peptide
  • Control Peptide - Blocking Peptide
  • PCA Assays
  • ImmunoBlot
This peptide sequence corresponds to a class I (Kb)-restricted peptide epitope of OVA, an octameric peptide from ovalbumin presented by the class I MHC molecule, H-2Kb.
Ovalbumin, 257-264 Control Peptide is suitable for use in ELISA, Western Blot, Dot blot, PCA, and other assays. Control peptide should be used at 1.0 µg per 1.0 µl of antiserum in per assay. Specific conditions for reactivity should be optimized by the end user.