C(Nyps)-Ovalbumin (323-339)
Supplier: Rockland Immunochemicals
000-001-M44
RL000-001-M44EA 199.54 USD
RL000-001-M44
C(Nyps)-Ovalbumin (323-339)
Proteins and Peptides
Ovalbumin Control Peptide
  • Control Peptide - Blocking Peptide
  • PCA Assays
  • ImmunoBlot
This peptide sequence corresponds to a class I (Kb)-restricted peptide epitope of OVA, an octameric peptide from ovalbumin presented by the class I MHC molecule, H-2Kb.
Ovalbumin, C-Nyps 323-339 Control Peptide is suitable for use in ELISA, Western Blot, Dot blot, PCA, and other assays. Control peptide should be used at 1.0 µg per 1.0 µl of antiserum in per assay. Specific conditions for reactivity should be optimized by the end user.