LL-37, 7-37
Supplier: Rockland Immunochemicals
000-001-M36
RL000-001-M36EA 265.31 USD
RL000-001-M36
LL-37, 7-37
Proteins and Peptides
Cathelicidin (LL-37) Control Peptide
  • Control Peptide - Blocking Peptide
  • PCA Assays
  • ImmunoBlot
Cathelicidin (LL-37) are a family of antimicrobial proteins found in lysosomes of macrophages and polymorphonuclear leukocytes. Along with the family of proteins known as defensins, cathelicidins participate in the first line of defense by preventing local infection and systemic invasion of microbes. LL-37 binds to bacterial lipopolysaccharides (LPS) and is a potent chemotactic factor for recruiting mast cells to sites of inflammation. LL-37 is present in inflammatory skin diseases that include psoriasis, sub-acute lupus erthematosus, dermatitis and nickel contact hypersensitivity. It is not found in normal skin epidermis. The secreted protein is expressed primarily in bone marrow, testis and neutrophils.
LL-37, 7-37 Control Peptide is suitable for use in ELISA, Western Blot, Dot blot, PCA, and other assays. Control peptide should be used at 1.0 µg per 1.0 µl of antiserum in per assay. Specific conditions for reactivity should be optimized by the end user.