Would you like to log in again?
Rat pTH 7-34, Rockland Immunochemicals
Supplier: Rockland Immunochemical
000-001-M32
RL000-001-M32EA 323.02 USD InStock
RL000-001-M32
Rat pTH 7-34, Rockland Immunochemicals
Proteins and Peptides
Parathyroid hormone Control Peptide
  • Control Peptide - Blocking Peptide
  • PCA Assays
  • ImmunoBlot
Parathyroid hormone (pTH) is the most important endocrine regulator of Ca2+ and phosphorus concentration in the extracellular fluid. pTH is secreted from cells of the parathyroid glands and acts for the most part via the binding to specific G-protein coupled receptors on the surface of  target cells in bone and kidney, utilizing the second messenger adenylate cyclase to regulate cAMP levels in cells.  Like most other protein hormones, parathyroid hormone is synthesized as a preprohormone and is processed intracellularly.  The mature hormone, 84 amino acids in length, is released from secretory vesicles into the blood.  Biologic effects of large C-terminal parathyroid hormone (PTH) fragments, such as pTH 7-84, are thought to regulate PTH secretion via an autocrine/paracrine regulatory mechanism
Parathyroid hormone (pTH) Rat 7-34 Control Peptide is suitable for use in ELISA, Western Blot, Dot blot, PCA, and other assays. Control peptide should be used at 1.0 µg per 1.0 µl of antiserum in per assay. Specific conditions for reactivity should be optimized by the end user.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty