Pool of 23 HLA class I restricted T cell epitopes from human cytomegalovirus, Epstein-Barr virus and influenza virus.
- Control Peptide - Blocking Peptide
- PCA Assays
- ImmunoBlot
Pool of 23 HLA class I restricted T cell epitopes from human cytomegalovirus, Epstein-Barr virus and influenza virus. Stimulates the release of IFN-gamma from CD8+ T cells in individuals with certain highly frequent HLA types (HLA: A1, A2, A3, A11, A24, A68, B7, B8, B27, B35, B44). Chosen epitopes are presented by the most common Caucasian HLA types, whose cumulative frequencies represent >100% of Caucasian individuals. Useful as a peptide specific positive control in ELISPOT, CTL and intracellular cytokine assays using human PBMC.
T-Cell Assay Panel Control Peptides are suitable for use in ELISA, Western Blot, Dot blot, PCA, and other assays. Control peptide should be used at 1.0 µg per 1.0 µl of antiserum in per assay. Specific conditions for reactivity should be optimized by the end user.