Would you like to log in again?
Hemoglobin C Control Peptide, Rockland Immunochemicals
Supplier: Rockland Immunochemical
000-001-GS7
76221-838EA 144.07 USD InStock
76221-838
Hemoglobin C Control Peptide, Rockland Immunochemicals
Proteins and Peptides
Hemoglobin C Control Peptide
  • Control Peptide - Blocking Peptide
  • PCA Assays
  • ImmunoBlot
HbC peptite corresponds to the E6K mutant in the amino-termianl portion of hemoglobin beta subunit. Functional hemoglobin (Hb) is a hetero tetramer composed of 2 alpha and 2 beta subunits (α2β2). Common isoform variants of hemoglobin include HbA, HbS, HbC, HbF, and HbA2. Sickle cell disease (SCD), thalassemias and hemoglobinopathies occur when aberrant forms of hemoglobin are expressed in children and adults. Globin gene mutations affect the structure and expression levels of Hb. Sickle cell disease and the more benign sickle cell trait are observed in more than 100 million people globally. Less significant than the SCD-E6V, HbC E6K mutation causes a mild hemolytic anemia. HbC peptide is suitable for use as a control with the HbC antibody. This peptide is ideal for investigators involved in Cardiovascular and developmental biology research.
HbC peptide can be used as a control with the HbC antibody. Control peptide should be used at 1.0 µg per 1.0 µl of antiserum in per assay.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty