Would you like to log in again?
MEK2 C-Term Blocking Peptide, Rockland Immunochemicals
Supplier: Rockland Immunochemical
000-001-GQ2
76222-534EA 144.07 USD InStock
76222-534
MEK2 C-Term Blocking Peptide, Rockland Immunochemicals
Proteins and Peptides
MEK2 C-Term Blocking Control Peptide
  • Control Peptide - Blocking Peptide
  • PCA Assays
  • ImmunoBlot
Mitogen-activated protein kinase kinase 2, also known as MEK2 or MKK2, is an integral component of the MAP kinase cascade that regulates cell growth and differentiation. This pathway also plays a key role in synaptic plasticity in the brain. Activated MEK 2 acts as a dual specificity kinase phosphorylating both a threonine and a tyrosine residue on MAP kinase. MEK1 and MEK2 are about 80% identical to each other, and nearly identical within the kinase domain. MEK2 is ideal for investigators involved in Neuroscience, Cell Signaling and Cancer Research.
MEK2 C-term blocking peptide is suitable as a control for polyclonal or monoclonal anti-MEK2 in immunological assays.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty