F-Box Protein Fbp5A Peptide
Supplier: Rockland Immunochemicals
000-001-490
RL000-001-490EA 151.2 USD
RL000-001-490
F-Box Protein Fbp5A Peptide
Proteins and Peptides
F-Box Protein Fbp5A Control Peptide
  • Control Peptide - Blocking Peptide
  • PCA Assays
  • ImmunoBlot
Fbp5A peptide for use in control assays with purified antibodies Anti-Fbp5A (p/n 600-401-490).
Control peptide should be used at 1.0 µg per 1.0 µl of antiserum in per assay.